Assay Details
Target Gene Details
Entrez Gene ID: | 77254 |
Gene Name: | Yip1 interacting factor homolog B (S. cerevisiae) |
Gene Aliases: |
9430029K10Rik, Yip1b |
Location: |
Chr.7:29238308-29247532 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yif1b | NM_001110201.1 | NP_001103671.1 | ||
| NM_029887.3 | NP_084163.2 | |||
| AK020444.1 | BAB32103.1 | |||
| BC109331.2 | AAI09332.1 | |||
| BU921944.1 | ||||
| BU953116.1 |

Genomic Map