Assay Details
Target Gene Details
Entrez Gene ID: | 56632 |
Gene Name: | sphingosine kinase 2 |
Gene Aliases: |
C76851 |
Location: |
Chr.7:45709463-45718002 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sphk2 | NM_001172561.1 | 1 | 237 | NP_001166032.1 |
| NM_203280.3 | 1 | 237 | NP_975009.1 | |
| AF245448.1 | 1 | 187 | AAF74125.1 | |
| AK146414.1 | 1 | 139 | ||
| AK151990.1 | 1 | 137 | BAE30855.1 | |
| BC053737.1 | 1 | 150 | AAH53737.1 | |
| BY256088.1 | 1 | 237 |
More Information
Set Membership: |
|

Genomic Map