Assay Details
Target Gene Details
Entrez Gene ID: | 56632 |
Gene Name: | sphingosine kinase 2 |
Gene Aliases: |
C76851 |
Location: |
Chr.7:45709463-45718002 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sphk2 | NM_001172561.1 | NP_001166032.1 | ||
| NM_020011.5 | NP_064395.2 | |||
| NM_203280.3 | NP_975009.1 | |||
| AF245448.1 | AAF74125.1 | |||
| AF415214.1 | AAL07500.1 | |||
| AK004951.1 | BAB23694.1 | |||
| AK146414.1 | ||||
| AK151990.1 | BAE30855.1 | |||
| AK154321.1 | BAE32512.1 | |||
| AK177087.1 | ||||
| BC006941.1 | AAH06941.1 | |||
| BC053737.1 | AAH53737.1 | |||
| BC092084.1 | AAH92084.1 |
More Information
Set Membership: |
|

Genomic Map