Assay Details
Target Gene Details
Entrez Gene ID: | 55925 |
Gene Name: | synaptotagmin VIII |
Gene Aliases: |
- |
Location: |
Chr.7:142434462-142440399 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syt8 | NM_001285857.1 | 5 | 609 | NP_001272786.1 |
| NM_001285858.1 | 5 | 620 | NP_001272787.1 | |
| NM_001285861.1 | 5 | 610 | NP_001272790.1 | |
| NM_018802.5 | 5 | 731 | NP_061272.2 | |
| AB026805.1 | 5 | 610 | BAA85777.1 | |
| AK075832.1 | 5 | 732 | BAC35993.1 | |
| BC100747.1 | 2 | 444 | AAI00748.1 | |
| BC100748.1 | 5 | 585 | AAI00749.1 | |
| BC103602.1 | 5 | 585 | AAI03603.1 | |
| BC103665.1 | 5 | 596 | AAI03666.1 | |
| U20107.1 | 3 | 391 | AAA87723.1 |
More Information
Set Membership: |
|

Genomic Map