Assay Details
Target Gene Details
Entrez Gene ID: | 12751 |
Gene Name: | tripeptidyl peptidase I |
Gene Aliases: |
Cln2, TPP-I |
Location: |
Chr.7:105744847-105752282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpp1 | NM_009906.5 | NP_034036.1 | ||
| AF111172.1 | AAD03083.1 | |||
| AJ011912.1 | CAA09863.1 | |||
| AK002418.1 | BAB22085.1 | |||
| AK048279.1 | BAC33293.1 | |||
| AK083832.1 | BAC39034.1 | |||
| AK146759.1 | ||||
| AK155021.1 | BAE32994.1 | |||
| AK165669.1 | ||||
| AK166648.1 | ||||
| AK169924.1 | BAE41462.1 | |||
| AK170781.1 | BAE42026.1 | |||
| AK212650.1 | ||||
| BC024820.1 | AAH24820.1 |
More Information
Set Membership: |
|

Genomic Map