Assay Details
Target Gene Details
Entrez Gene ID: | 12751 |
Gene Name: | tripeptidyl peptidase I |
Gene Aliases: |
Cln2, TPP-I |
Location: |
Chr.7:105744847-105752282 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpp1 | NM_009906.5 | 6 | 573 | NP_034036.1 |
| AF111172.1 | 6 | 581 | AAD03083.1 | |
| AJ011912.1 | 6 | 567 | CAA09863.1 | |
| AK002418.1 | 6 | 592 | BAB22085.1 | |
| AK048279.1 | 6 | 574 | BAC33293.1 | |
| AK083832.1 | 5 | 1838 | BAC39034.1 | |
| AK146759.1 | 6 | 566 | ||
| AK155021.1 | 6 | 592 | BAE32994.1 | |
| AK165669.1 | 6 | 598 | ||
| AK166648.1 | 4 | 367 | ||
| AK169924.1 | 6 | 580 | BAE41462.1 | |
| AK170781.1 | 6 | 569 | BAE42026.1 | |
| AK212650.1 | ||||
| BC024820.1 | 6 | 575 | AAH24820.1 |
More Information
Set Membership: |
|

Genomic Map