Assay Details
Target Gene Details
Entrez Gene ID: | 67091 |
Gene Name: | trafficking protein particle complex 6A |
Gene Aliases: |
1810073E21Rik, 4930519D19Rik, AI480686, TRS33, mhyp |
Location: |
Chr.7:19507406-19516145 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc6a | NM_025960.3 | NP_080236.2 | ||
| AK007965.1 | BAB25376.1 | |||
| AK008022.1 | BAB25415.1 | |||
| AK008462.2 | BAB25681.2 | |||
| AK088832.1 | BAC40600.1 | |||
| BC020128.1 | ||||
| BC037154.1 | AAH37154.1 | |||
| BC106114.1 | AAI06115.1 |

Genomic Map