Assay Details
Target Gene Details
Entrez Gene ID: | 58206 |
Gene Name: | zinc finger and BTB domain containing 32 |
Gene Aliases: |
4930524C15Rik, FAXF, FAZF, PLZP, Rog, Tzfp |
Location: |
Chr.7:30589680-30602818 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb32 | NM_021397.2 | NP_067372.2 | ||
| AF241232.1 | AAF61244.1 | |||
| AK015881.1 | BAB30015.1 | |||
| BC137687.1 |
More Information
Set Membership: |
|

Genomic Map