Assay Details
Target Gene Details
Entrez Gene ID: | 72723 |
Gene Name: | zinc finger protein 74 |
Gene Aliases: |
2810054M15Rik, KRAB8, Zfp66, Znf569, mszf21, mszf77 |
Location: |
Chr.7:29931711-29955843 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp74 | NM_178384.3 | NP_848471.2 | ||
| BC050048.1 | AAH50048.1 |
More Information
Set Membership: |
|

Genomic Map