Assay Details
Target Gene Details
Entrez Gene ID: | 20909 |
Gene Name: | syntaxin 4A (placental) |
Gene Aliases: |
Stx4, Syn-4, Syn4 |
Location: |
Chr.7:127841769-127849686 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Stx4a | NM_009294.3 | NP_033320.1 | ||
| AK145909.1 | BAE26743.1 | |||
| AK161971.1 | BAE36660.1 | |||
| BC005791.1 | AAH05791.1 | |||
| BC011491.1 | AAH11491.1 | |||
| BC052023.1 | AAH52023.1 | |||
| U76832.1 | AAB18991.1 |
More Information
Set Membership: |
|

Genomic Map