Assay Details
Target Gene Details
Entrez Gene ID: | 20887 |
Gene Name: | sulfotransferase family 1A, phenol-preferring, member 1 |
Gene Aliases: |
AI266890, PST, ST1A1, Stp, Stp1 |
Location: |
Chr.7:126672865-126676483 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sult1a1 | NM_133670.1 | NP_598431.1 | ||
| AK002700.1 | ||||
| BC005413.1 | AAH05413.1 | |||
| L02331.1 |
More Information
Set Membership: |
|

Genomic Map