Assay Details
Target Gene Details
Entrez Gene ID: | 21803 |
Gene Name: | transforming growth factor, beta 1 |
Gene Aliases: |
TGF-beta1, TGFbeta1, Tgfb, Tgfb-1 |
Location: |
Chr.7:25687002-25704996 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb1 | NM_011577.1 | 7 | 1986 | NP_035707.1 |
| AJ009862.1 | 7 | 1986 | CAA08900.1 | |
| AK144163.1 | 7 | 1988 | BAE25742.1 | |
| BC013738.1 | 7 | 1433 | AAH13738.1 | |
| M13177.1 | 7 | 1471 | AAA40423.1 |
More Information
Set Membership: |
|

Genomic Map