Assay Details
Target Gene Details
Entrez Gene ID: | 102465638 |
Gene Name: | microRNA 7055 |
Gene Aliases: |
mmu-mir-7055 |
Location: |
Chr.7:45173303-45173361 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir7055 | NR_106022.1 |
Target Gene Details
Entrez Gene ID: | 72961 |
Gene Name: | solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7 |
Gene Aliases: |
2900052E22Rik, AI851913, Vglut1 |
Location: |
Chr.7:45163921-45176139 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc17a7 | NM_182993.2 | NP_892038.2 | ||
| AK159064.1 | BAE34790.1 | |||
| BC028989.1 | AAH28989.1 | |||
| BC054462.1 | AAH54462.1 |
More Information
Set Membership: |
|

Genomic Map