Assay Details
Target Gene Details
Entrez Gene ID: | 21804 |
Gene Name: | transforming growth factor beta 1 induced transcript 1 |
Gene Aliases: |
ARA55, Hic5, TSC-5, hic-5 |
Location: |
Chr.7:128246799-128255699 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb1i1 | NM_001289550.1 | NP_001276479.1 | ||
| NM_001289551.1 | NP_001276480.1 | |||
| NM_001289552.1 | NP_001276481.1 | |||
| NM_001289553.1 | NP_001276482.1 | |||
| AK003170.1 | ||||
| AK041233.1 | ||||
| AK085042.1 | ||||
| AK087114.1 | ||||
| AK156423.1 | BAE33707.1 | |||
| AK158409.1 | BAE34493.1 | |||
| BC002049.1 | AAH02049.1 | |||
| BC056362.1 | AAH56362.1 | |||
| DQ143891.1 | AAZ82195.1 | |||
| DQ143892.1 | AAZ82196.1 | |||
| DQ143893.1 | AAZ82197.1 | |||
| DQ143894.1 | AAZ82198.1 | |||
| DQ143895.1 | AAZ82199.1 | |||
| DQ143896.1 | AAZ82200.1 | |||
| DQ143897.1 | AAZ82201.1 | |||
| DQ143898.1 | AAZ82202.1 | |||
| DQ143899.1 | AAZ82203.1 | |||
| DQ143900.1 | AAZ82204.1 | |||
| L22482.1 | AAA62226.1 |
More Information
Set Membership: |
|

Genomic Map