Assay Details
Target Gene Details
Entrez Gene ID: | 212518 |
Gene Name: | shadow of prion protein |
Gene Aliases: |
C630041J07 |
Location: |
Chr.7:140150628-140154659 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sprn | NM_183147.2 | NP_898970.1 | ||
| AK049995.1 | BAC34022.1 | |||
| BC056484.1 | AAH56484.1 |

Genomic Map