Assay Details
Target Gene Details
Entrez Gene ID: | 269954 |
Gene Name: | tubulin tyrosine ligase-like family, member 13 |
Gene Aliases: |
1700111A04Rik, 4921538O11, BC024578 |
Location: |
Chr.7:80246376-80260821 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ttll13 | NM_177765.3 | 11 | 1788 | NP_808433.2 |
| AK029565.1 | 11 | 1787 | BAC26514.1 | |
| AK161519.1 | 11 | 1611 | ||
| AM690757.1 | 10 | 1489 | ||
| BC145749.1 | 10 | 1396 |
More Information
Set Membership: |
|

Genomic Map