Assay Details
Target Gene Details
Entrez Gene ID: | 26459 |
Gene Name: | solute carrier family 27 (fatty acid transporter), member 5 |
Gene Aliases: |
FACVL3, FATP5, VLCS-H2, VLCSH2, Vlacsr |
Location: |
Chr.7:12988346-12998192 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc27a5 | NM_009512.2 | 7 | 1740 | NP_033538.2 |
| AF072760.1 | 7 | 1529 | AAC40189.1 | |
| AI173562.1 | 4 | 597 | ||
| AJ223959.1 | 7 | 1698 | CAA11688.1 | |
| AK144301.1 | BAE25821.1 | |||
| BC013272.1 | 7 | 1705 | AAH13272.1 | |
| BC013335.1 | 7 | 1705 | AAH13335.1 | |
| BC115421.2 | 7 | 1680 | ||
| BC145823.1 | 7 | 1680 | ||
| BC145825.1 | 7 | 1680 |
More Information
Set Membership: |
|

Genomic Map