Assay Details
Target Gene Details
Entrez Gene ID: | 497652 |
Gene Name: | adrenocortical dysplasia |
Gene Aliases: |
- |
Location: |
Chr.8:105698157-105701114 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acd | NM_001012638.1 | 7 | 597 | NP_001012656.1 |
| AK032683.1 | 7 | 577 | BAE43282.1 | |
| AK145959.1 | 7 | 694 | BAE26785.1 | |
| AK160599.1 | 7 | 668 | BAE35904.1 | |
| AK170606.1 | 6 | 578 | ||
| AK171345.1 | BAE42403.1 | |||
| AK171458.1 | 7 | 588 | ||
| BC004682.1 | 1 | 124 | AAH04682.1 | |
| BC138694.1 | 7 | 562 | ||
| BC138695.1 | 7 | 562 |

Genomic Map