Assay Details
Target Gene Details
Entrez Gene ID: | 11440 |
Gene Name: | cholinergic receptor, nicotinic, alpha polypeptide 6 |
Gene Aliases: |
Acra6, Nica6 |
Location: |
Chr.8:27403212-27413944 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrna6 | NM_021369.2 | NP_067344.2 | ||
| AJ245706.1 | CAB53472.1 | |||
| AK149262.1 | BAE28777.1 | |||
| AY574265.1 | AAS90361.1 | |||
| BC031985.1 | AAH31985.1 |
More Information
Set Membership: |
|

Genomic Map