Assay Details
Target Gene Details
Entrez Gene ID: | 12857 |
Gene Name: | cytochrome c oxidase subunit IV isoform 1 |
Gene Aliases: |
AL024441, COX, COX IV-1, COXIV, Cox4, Cox4a, IV-1 |
Location: |
Chr.8:120668225-120674209 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox4i1 | NM_009941.3 | NP_034071.2 | ||
| BY250039.1 |
More Information
Set Membership: |
|

Genomic Map