Assay Details
Target Gene Details
Entrez Gene ID: | 12857 |
Gene Name: | cytochrome c oxidase subunit IV isoform 1 |
Gene Aliases: |
AL024441, COX, COX IV-1, COXIV, Cox4, Cox4a, IV-1 |
Location: |
Chr.8:120668225-120674209 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox4i1 | NM_001293559.1 | NP_001280488.1 | ||
| NM_009941.3 | NP_034071.2 | |||
| AK012583.1 | BAB28333.1 | |||
| AK019276.1 | BAB31643.1 | |||
| AK150447.1 | BAE29569.1 | |||
| AV069678.1 | ||||
| AV469576.1 | ||||
| BC132269.1 | ||||
| BC132275.1 | ||||
| BE992841.1 | ||||
| BY250039.1 | ||||
| M37829.1 | AAB02140.1 | |||
| X54691.1 | CAA38507.1 |
More Information
Set Membership: |
|

Genomic Map