Assay Details
Target Gene Details
Entrez Gene ID: | 12857 |
Gene Name: | cytochrome c oxidase subunit IV isoform 1 |
Gene Aliases: |
AL024441, COX, COX IV-1, COXIV, Cox4, Cox4a, IV-1 |
Location: |
Chr.8:120668225-120674209 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox4i1 | NM_001293559.1 | 5 | 588 | NP_001280488.1 |
| NM_009941.3 | 5 | 663 | NP_034071.2 | |
| AK012583.1 | 5 | 600 | BAB28333.1 | |
| AK019276.1 | 5 | 565 | BAB31643.1 | |
| AK150447.1 | 5 | 593 | BAE29569.1 | |
| AV069678.1 | 2 | 216 | ||
| BC132269.1 | 5 | 584 | ||
| BC132275.1 | 5 | 584 | ||
| BE992841.1 | 1 | 100 | ||
| M37829.1 | 5 | 586 | AAB02140.1 | |
| X54691.1 | 5 | 588 | CAA38507.1 |
More Information
Set Membership: |
|

Genomic Map