Assay Details
Target Gene Details
Entrez Gene ID: | 18117 |
Gene Name: | ER membrane protein complex subunit 8 |
Gene Aliases: |
Cox4nb, Fam158b, Noc4 |
Location: |
Chr.8:120653914-120668112 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Emc8 | NM_010926.5 | NP_035056.1 | ||
| AF052621.1 | AAC12933.1 | |||
| AK042704.1 | BAC31338.1 | |||
| AK076710.1 | ||||
| AK143131.1 | BAE25280.1 | |||
| AK148190.1 | ||||
| AK161438.1 | BAE36396.1 | |||
| AK164130.1 | BAE37640.1 | |||
| AK201805.1 | ||||
| BC009103.1 | AAH09103.1 |

Genomic Map