Assay Details
Target Gene Details
Entrez Gene ID: | 11459 |
Gene Name: | actin, alpha 1, skeletal muscle |
Gene Aliases: |
AA959943, Acta-2, Acts, Actsk-1 |
Location: |
Chr.8:123891758-123894775 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acta1 | NM_001272041.1 | NP_001258970.1 | ||
| NM_009606.3 | NP_033736.1 | |||
| AK027919.1 | ||||
| BC014877.1 | AAH14877.1 | |||
| M12233.1 | AAA37143.1 | |||
| M12866.1 | AAA37164.1 | |||
| X03766.1 | CAA27397.1 |
More Information
Set Membership: |
|

Genomic Map