Assay Details
Target Gene Details
Entrez Gene ID: | 71955 |
Gene Name: | increased sodium tolerance 1 homolog (yeast) |
Gene Aliases: |
2400003C14Rik, AW536298, mKIAA0174 |
Location: |
Chr.8:109671321-109693294 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ist1 | NM_028018.2 | NP_082294.1 | ||
| AK010262.1 | BAB26804.1 | |||
| AK050468.1 | BAC34273.1 | |||
| AK088192.1 | BAC40200.1 | |||
| AK122215.1 | 5 | 2819 | BAC65497.1 | |
| AK152830.1 | BAE31530.1 | |||
| AK186831.1 | ||||
| AK204355.1 | ||||
| AK206037.1 | ||||
| AK207621.1 | ||||
| AK215347.1 | ||||
| AK218427.1 | ||||
| BC017605.1 | AAH17605.1 | |||
| BC039052.1 | AAH39052.1 |
More Information
Set Membership: |
|

Genomic Map