Assay Details
Target Gene Details
Entrez Gene ID: | 234854 |
Gene Name: | cyclin-dependent kinase 10 |
Gene Aliases: |
BC017131 |
Location: |
Chr.8:123224840-123232256 on Build GRCm38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk10 | NM_194444.2 | NP_919426.2 | ||
| NM_194446.2 | NP_919428.1 | |||
| AK133918.1 | BAE21925.1 | |||
| AK144801.1 | BAE26074.1 | |||
| AK170594.1 | ||||
| BC017131.1 | ||||
| BC023736.1 | ||||
| DQ288857.1 | ABB96224.1 |
More Information
Set Membership: |
|

Genomic Map