Assay Details
Target Gene Details
Entrez Gene ID: | 69550 |
Gene Name: | bone marrow stromal cell antigen 2 |
Gene Aliases: |
2310015I10Rik, Bst-2, C87040, CD317, DAMP-1, GREG |
Location: |
Chr.8:71534259-71537437 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bst2 | NM_198095.2 | 4 | 489 | NP_932763.1 |
| AK009361.1 | 4 | 289 | BAC25254.1 | |
| AK154399.1 | 4 | 482 | BAE32560.1 | |
| BC027328.1 | 4 | 523 | AAH27328.1 | |
| BC056638.1 | 4 | 496 | AAH56638.1 | |
| BC087949.1 | 4 | 481 | AAH87949.1 |

Genomic Map