Assay Details
Target Gene Details
Entrez Gene ID: | 68251 |
Gene Name: | BRISC and BRCA1 A complex member 1 |
Gene Aliases: |
5430437P03Rik, Merit40 |
Location: |
Chr.8:71396844-71404772 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Babam1 | NM_026636.2 | NP_080912.2 | ||
| AK077402.1 | BAC36785.1 | |||
| AK147083.1 | BAE27663.1 | |||
| AK183167.1 | ||||
| AK210692.1 | ||||
| BC005692.1 | AAH05692.1 |

Genomic Map