Assay Details
Target Gene Details
Entrez Gene ID: | 73609 |
Gene Name: | RIKEN cDNA 1700125H03 gene |
Gene Aliases: |
4930510D22Rik |
Location: |
Chr.8:68368459-68375710 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700125H03Rik | NR_038181.1 | |||
| AK015741.1 |
Target Gene Details
Entrez Gene ID: | 234356 |
Gene Name: | chondroitin sulfate N-acetylgalactosaminyltransferase 1 |
Gene Aliases: |
4732435N03Rik, CSGalNAcT-1 |
Location: |
Chr.8:68356781-68735673 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csgalnact1 | NM_001252623.1 | NP_001239552.1 | ||
| NM_172753.5 | NP_766341.4 | |||
| AK028688.1 | BAC26066.1 | |||
| AK033522.1 | BAC28340.1 | |||
| AK080687.1 | BAC37982.1 | |||
| AK133567.1 | BAE21729.1 | |||
| AK144089.1 | BAE25692.1 | |||
| BC026599.1 | AAH26599.1 | |||
| BC089162.1 | AAH89162.1 |
More Information
Set Membership: |
|

Genomic Map