Assay Details
Target Gene Details
Entrez Gene ID: | 67687 |
Gene Name: | RIKEN cDNA 1700011L22 gene |
Gene Aliases: |
- |
Location: |
Chr.8:79210423-79248897 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700011L22Rik | NM_026315.1 | NP_080591.1 | ||
| AK005876.1 | BAB24292.1 |

Genomic Map