Assay Details
Target Gene Details
Entrez Gene ID: | 11556 |
Gene Name: | adrenergic receptor, beta 3 |
Gene Aliases: |
Adrb-3 |
Location: |
Chr.8:27225776-27230805 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adrb3 | NM_013462.3 | NP_038490.2 | ||
| AF193027.1 | AAF05768.1 | |||
| AK136495.1 | BAE23009.1 | |||
| AK143775.1 | BAE25534.1 | |||
| BC132000.1 | ||||
| S53290.1 | AAB24836.1 |
More Information
Set Membership: |
|

Genomic Map