Assay Details
Target Gene Details
Entrez Gene ID: | 382051 |
Gene Name: | pyruvate dehyrogenase phosphatase catalytic subunit 2 |
Gene Aliases: |
4833426J09Rik, Gm1705, mKIAA1348 |
Location: |
Chr.8:104591468-104599026 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdp2 | NM_001024606.1 | NP_001019777.1 | ||
| BC094946.1 | AAH94946.1 | |||
| BY017072.1 |
More Information
Set Membership: |
|

Genomic Map