Assay Details
Target Gene Details
Entrez Gene ID: | 11497 |
Gene Name: | a disintegrin and metallopeptidase domain 3 (cyritestin) |
Gene Aliases: |
Cyrn1, Taz83, tMDC |
Location: |
Chr.8:24677232-24726044 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam3 | NM_009619.4 | NP_033749.2 | ||
| BC049609.1 | AAH49609.1 | |||
| BC060975.1 | AAH60975.1 | |||
| X64227.1 | CAA45534.1 |
More Information
Set Membership: |
|

Genomic Map