Assay Details
Target Gene Details
Entrez Gene ID: | 64933 |
Gene Name: | adaptor-related protein complex 3, mu 2 subunit |
Gene Aliases: |
5830445E16Rik, AP-3B |
Location: |
Chr.8:22787354-22805654 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap3m2 | NM_001122820.1 | 3 | 564 | NP_001116292.1 |
| NM_029505.3 | 3 | 502 | NP_083781.2 | |
| AK017991.1 | 3 | 451 | ||
| AK077365.1 | 3 | 479 | BAC36770.1 | |
| AK080668.1 | 3 | 470 | ||
| AK081220.1 | 3 | 471 | BAC38169.1 | |
| AK082988.1 | 3 | 460 | BAC38723.1 | |
| AK134634.1 | 3 | 514 | BAE22219.1 | |
| AY039763.1 | 3 | 482 | AAK73278.1 | |
| BB610934.1 | 3 | 502 | ||
| BC027301.1 | 3 | 448 | AAH27301.1 | |
| BC030484.1 | 3 | 486 | AAH30484.1 |
More Information
Set Membership: |
|

Genomic Map