Assay Details
Target Gene Details
Entrez Gene ID: | 12745 |
Gene Name: | calmegin |
Gene Aliases: |
4930459O04Rik, AI528775, Cln |
Location: |
Chr.8:83389826-83428552 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clgn | NM_009904.2 | NP_034034.2 | ||
| AK019534.1 | BAB31782.1 | |||
| AK034525.1 | ||||
| BC050767.1 | AAH50767.1 | |||
| D14117.1 | BAA03180.1 | |||
| D86323.1 | BAA22591.1 | |||
| U08373.1 | AAA20599.1 |
More Information
Set Membership: |
|

Genomic Map