Assay Details
Target Gene Details
Entrez Gene ID: | 11502 |
Gene Name: | a disintegrin and metallopeptidase domain 9 (meltrin gamma) |
Gene Aliases: |
AU020942, MDC9, Mltng, mKIAA0021 |
Location: |
Chr.8:24949611-25016933 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam9 | NM_001270996.1 | 1 | 164 | NP_001257925.1 |
| NM_007404.2 | 1 | 164 | NP_031430.2 | |
| AK122188.1 | 1 | 147 | BAC65470.1 | |
| AK148174.1 | 1 | 50 | BAE28394.1 | |
| AK155582.1 | 1 | 125 | ||
| AK155672.1 | 1 | 57 | BAE33380.1 | |
| AK161778.1 | 1 | 88 | ||
| BC047156.1 | 1 | 42 | AAH47156.1 | |
| BY780211.1 | 1 | 108 | ||
| CJ078757.1 | 1 | 169 | ||
| U41765.1 | AAC52446.1 |
More Information
Set Membership: |
|

Genomic Map