Assay Details
Target Gene Details
Entrez Gene ID: | 11502 |
Gene Name: | a disintegrin and metallopeptidase domain 9 (meltrin gamma) |
Gene Aliases: |
AU020942, MDC9, Mltng, mKIAA0021 |
Location: |
Chr.8:24949611-25016933 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam9 | NM_001270996.1 | 11 | 1207 | NP_001257925.1 |
| NM_007404.2 | 11 | 1207 | NP_031430.2 | |
| AK122188.1 | 11 | 1190 | BAC65470.1 | |
| AK148174.1 | 11 | 1093 | BAE28394.1 | |
| AK155582.1 | 11 | 1168 | ||
| AK155672.1 | 11 | 1100 | BAE33380.1 | |
| AK161778.1 | 11 | 1131 | ||
| BC047156.1 | 11 | 1085 | AAH47156.1 | |
| U41765.1 | 11 | 1079 | AAC52446.1 |
More Information
Set Membership: |
|

Genomic Map