Assay Details
Target Gene Details
Entrez Gene ID: | 64933 |
Gene Name: | adaptor-related protein complex 3, mu 2 subunit |
Gene Aliases: |
5830445E16Rik, AP-3B |
Location: |
Chr.8:22787354-22805654 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap3m2 | NM_001122820.1 | 7 | 1080 | NP_001116292.1 |
| NM_029505.3 | 7 | 1018 | NP_083781.2 | |
| AK077365.1 | 7 | 995 | BAC36770.1 | |
| AK081220.1 | 7 | 987 | BAC38169.1 | |
| AK082988.1 | 7 | 976 | BAC38723.1 | |
| AK134634.1 | 7 | 1030 | BAE22219.1 | |
| AK162771.1 | 2 | 123 | BAE37054.1 | |
| AK197365.1 | 1 | 81 | ||
| AY039763.1 | 7 | 998 | AAK73278.1 | |
| BC027301.1 | 7 | 964 | AAH27301.1 | |
| BC030484.1 | 7 | 1002 | AAH30484.1 |
More Information
Set Membership: |
|

Genomic Map