Assay Details
Target Gene Details
Entrez Gene ID: | 64933 |
Gene Name: | adaptor-related protein complex 3, mu 2 subunit |
Gene Aliases: |
5830445E16Rik, AP-3B |
Location: |
Chr.8:22787354-22805654 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap3m2 | NM_001122820.1 | NP_001116292.1 | ||
| NM_029505.3 | NP_083781.2 | |||
| AK077365.1 | BAC36770.1 | |||
| AK081220.1 | BAC38169.1 | |||
| AK082988.1 | BAC38723.1 | |||
| AK134634.1 | BAE22219.1 | |||
| AK162771.1 | BAE37054.1 | |||
| AY039763.1 | AAK73278.1 | |||
| BC027301.1 | AAH27301.1 | |||
| BC030484.1 | AAH30484.1 |
More Information
Set Membership: |
|

Genomic Map