Assay Details
Target Gene Details
Entrez Gene ID: | 234669 |
Gene Name: | carboxyesterase 2B |
Gene Aliases: |
- |
Location: |
Chr.8:104828494-104840093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces2b | NM_198171.2 | NP_937814.1 | ||
| BC058815.1 | AAH58815.1 |
More Information
Set Membership: |
|

Genomic Map