Assay Details
Target Gene Details
Entrez Gene ID: | 23808 |
Gene Name: | ash2 (absent, small, or homeotic)-like (Drosophila) |
Gene Aliases: |
- |
Location: |
Chr.8:25816000-25847694 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ash2l | NM_001080793.2 | NP_001074262.1 | ||
| NM_001286207.1 | NP_001273136.1 | |||
| NM_011791.3 | NP_035921.2 | |||
| AB020983.2 | BAA35128.1 | |||
| AK087934.1 | BAC40047.1 | |||
| AK132471.1 | ||||
| AK145576.1 | BAE26521.1 | |||
| AK145790.1 | BAE26652.1 | |||
| AK145967.1 | BAE26792.1 | |||
| AK146254.1 | BAE27016.1 | |||
| AK146599.1 | BAE27293.1 | |||
| AK146756.1 | BAE27411.1 | |||
| AK146938.1 | BAE27547.1 | |||
| BC012957.1 | AAH12957.1 | |||
| BY266419.1 |
More Information
Set Membership: |
|

Genomic Map