Assay Details
Target Gene Details
Entrez Gene ID: | 668110 |
Gene Name: | synaptonemal complex central element protein 1 like |
Gene Aliases: |
4930481F22Rik, MMRP, PSESS, mmrp2 |
Location: |
Chr.8:113643213-113655533 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syce1l | NM_001048145.1 | NP_001041610.1 | ||
| NM_001286494.1 | NP_001273423.1 | |||
| NM_029139.1 | NP_083415.1 | |||
| AK015606.1 | BAB29903.1 | |||
| AY899935.1 | AAX18325.1 | |||
| AY899936.1 | AAX18326.1 | |||
| BC100478.1 | AAI00479.1 | |||
| BC141326.1 | ||||
| BC141347.1 |

Genomic Map