Assay Details
Target Gene Details
Entrez Gene ID: | 50770 |
Gene Name: | ATPase, class VI, type 11A |
Gene Aliases: |
4930558F19Rik, AU040868, Atpc1h, Ih |
Location: |
Chr.8:12757016-12868728 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp11a | NM_001293667.1 | NP_001280596.1 | ||
| NM_001293668.1 | NP_001280597.1 | |||
| NM_015804.3 | NP_056619.1 | |||
| NR_121603.1 | ||||
| AF156551.1 | AAF09449.1 | |||
| AK147497.1 | ||||
| AK173088.1 | ||||
| BC138715.1 |
Target Gene Details
Entrez Gene ID: | 100504249 |
Gene Name: | predicted gene 15350 |
Gene Aliases: |
OTTMUSG00000020611 |
Location: |
Chr.8:12828338-12831919 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15350 | NR_045775.1 | |||
| AK006628.1 | ||||
| AK018911.1 |
More Information
Set Membership: |
|

Genomic Map