Assay Details
Target Gene Details
Entrez Gene ID: | 72361 |
Gene Name: | carboxylesterase 2G |
Gene Aliases: |
2210023G05Rik, Ces2l |
Location: |
Chr.8:104961611-104969537 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces2g | NM_197999.2 | NP_932116.2 | ||
| AK146261.1 | ||||
| BC027185.1 | AAH27185.1 |
More Information
Set Membership: |
|

Genomic Map