Assay Details
Target Gene Details
Entrez Gene ID: | 66164 |
Gene Name: | nuclear import 7 homolog (S. cerevisiae) |
Gene Aliases: |
1110017C15Rik, 6330509M23Rik, AA408773, AA410017, PEachy |
Location: |
Chr.8:107056877-107060931 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nip7 | NM_001164472.1 | NP_001157944.1 | ||
| NM_025391.2 | NP_079667.2 | |||
| NR_028367.1 | ||||
| AK003739.1 | BAB22972.1 | |||
| AK006057.1 | ||||
| AK014290.1 | BAB29247.1 | |||
| BC003972.1 | AAH03972.1 | |||
| BE623972.1 |
More Information
Set Membership: |
|

Genomic Map