Assay Details
Target Gene Details
Entrez Gene ID: | 67443 |
Gene Name: | microtubule-associated protein 1 light chain 3 beta |
Gene Aliases: |
1010001C15Rik, Atg8, LC3b, Map1lc3 |
Location: |
Chr.8:121590468-121598047 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map1lc3b | NM_026160.4 | NP_080436.1 | ||
| AF255953.1 | AAL83723.1 | |||
| AK002795.1 | BAB22364.1 | |||
| AK003106.1 | BAB22569.1 | |||
| AK003205.1 | BAB22641.1 | |||
| AK003558.1 | BAB22855.1 | |||
| AK012604.1 | BAB28350.1 | |||
| AK151614.1 | BAE30551.1 | |||
| BC068180.1 | AAH68180.1 | |||
| BC110308.1 | AAI10309.1 |
More Information
Set Membership: |
|

Genomic Map