Assay Details
Target Gene Details
Entrez Gene ID: | 75687 |
Gene Name: | family with sequence similarity 65, member A |
Gene Aliases: |
2310066E14Rik, AI837433 |
Location: |
Chr.8:105594087-105622219 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam65a | NM_001081241.2 | NP_001074710.2 | ||
| AK052316.1 | ||||
| AK122573.1 | BAC65855.1 | |||
| AK184855.1 | ||||
| AK186816.1 | ||||
| BC079880.1 | AAH79880.1 |
Target Gene Details
Entrez Gene ID: | 100316712 |
Gene Name: | microRNA 1966 |
Gene Aliases: |
mir-1966, mmu-mir-1966 |
Location: |
Chr.8:105615466-105615573 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir1966 | NR_035492.1 |
More Information
Set Membership: |
|

Genomic Map