Assay Details
Target Gene Details
Entrez Gene ID: | 13909 |
Gene Name: | carboxylesterase 3B |
Gene Aliases: |
Ces31L, Ces3L, EG13909, ES31L, Es31, Gm4738 |
Location: |
Chr.8:105083730-105093929 on Build GRCm38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces3b | NM_001159415.1 | NP_001152887.1 | ||
| NM_144511.2 | NP_653094.2 | |||
| AI173514.1 | ||||
| BC010812.1 | AAH10812.1 | |||
| BC019147.1 | AAH19147.1 |
More Information
Set Membership: |
|

Genomic Map