Assay Details
Target Gene Details
Entrez Gene ID: | 14272 |
Gene Name: | farnesyltransferase, CAAX box, alpha |
Gene Aliases: |
FTA |
Location: |
Chr.8:25998722-26015601 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fnta | NM_008033.3 | 2 | 271 | NP_032059.1 |
| AK010744.1 | 2 | 272 | BAB27154.1 | |
| AK090346.1 | 2 | 271 | BAC41179.1 | |
| AK153252.1 | 2 | 266 | BAE31843.1 | |
| AK183567.1 | 2 | 111 | ||
| BC012711.1 | 2 | 289 | AAH12711.1 | |
| D49744.1 | 2 | 265 | BAA08578.1 |
More Information
Set Membership: |
|

Genomic Map