Assay Details
Target Gene Details
Entrez Gene ID: | 78833 |
Gene Name: | GINS complex subunit 3 (Psf3 homolog) |
Gene Aliases: |
2700085M18Rik, AI616142 |
Location: |
Chr.8:95633559-95645059 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gins3 | NM_030198.3 | 2 | 346 | NP_084474.1 |
| AK019225.1 | 2 | 347 | BAB31610.1 | |
| AK146574.1 | 2 | 377 | BAE27270.1 | |
| AK150602.1 | 2 | 378 | BAE29694.1 | |
| AK151243.1 | 2 | 378 | BAE30235.1 | |
| AK152572.1 | 2 | 378 | BAE31324.1 | |
| AK160606.1 | 2 | 347 | BAE35909.1 | |
| BC039794.1 | 2 | 343 | AAH39794.1 | |
| BC064746.1 | 2 | 335 | AAH64746.1 |

Genomic Map